m-Tolylacetat ≥97%

Lieferant: Thermo Scientific

L14602.14 L14602.22
L14602.14EA 34.5 EUR
L14602.14 L14602.22
m-Tolylacetat ≥97%
m-Tolylacetat

Purity: 97% CAS No.: 122-46-3 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 95°(203°F) Refractive Index: 1.5020 Boiling Point: 100°/14mm Beilstein Reference: 1862796 Merck Reference: 14,2583 MDL No.: MFCD00041910

Formel: C₉H₁₀O₂
Molecular Weight: 150,18 g/mol
Siedepunkt: 100 °C (14 mmHg)
Dichte: 1,050
Flammpunkt: 95 °C (203 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00041910
CAS-Nummer: 122-46-3
EINECS: 204-546-2
Merck Index: 14,2583

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR