4-Brom-2,6-dichlor-3-methylanilin

Lieferant: COMBI-BLOCKS

AN-1179-5G AN-1179-1G
COBBAN-1179-5GEA 595 EUR
COBBAN-1179-5G COBBAN-1179-1G
4-Brom-2,6-dichlor-3-methylanilin
4-Brom-2,6-dichlor-3-methylanilin
Formel: C₇H₆BrCl₂N
Molecular Weight: 254,9400 g/mol
MDL: MFCD09800852
CAS-Nummer: 62406-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR