Ethyl-4,4,4-trifluorcrotonat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl (E)-4,4,4-trifluorobut-2-enoate, Ethyl 4,4,4-trifluorocrotonate
001456-5G
001456-25G
FLUO001456-5GEA
51.8
EUR
FLUO001456-5G
FLUO001456-25G
Ethyl-4,4,4-trifluorcrotonat
Ethyl-4,4,4-trifluorcrotonat
Formel:
C₆H₇F₃O₂ Molecular Weight: 168,12 g/mol |
MDL:
MFCD00009903 CAS-Nummer: 25597-16-4 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...