Methyl-4,4,4-trifluorcrotonat ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
L11466.03
L11466.06
L11466.03EA
49.8
EUR
L11466.03
L11466.06
Methyl-4,4,4-trifluorcrotonat ≥97%
Methyl-4,4,4-trifluorcrotonat
Purity: 97%
CAS No.: 85694-31-1
Molecular Formula: CF3CH=CHCO2CH3
Formula Weight: 154.10
Refractive Index: 1.3535
Boiling Point: 94-95°
UN No.: 3272
MDL No.: MFCD00077567
Formel:
C₅H₅F₃O₂ Molecular Weight: 154,10 g/mol Siedepunkt: 94…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077567 CAS-Nummer: 85694-31-1 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...