Methyl-2-amino-5-methoxybenzoat 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 2-amino-5-methoxybenzoate

H54704.MD H54704.03 H54704.06
H54704.MDEA 69.6 EUR
H54704.MD H54704.03 H54704.06
Methyl-2-amino-5-methoxybenzoat 96%
Methyl-2-amino-5-methoxybenzoat

Purity: 96% CAS No.: 2475-80-1 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Melting Point: 32-33° MDL No.: MFCD09038482

Formel: C₉H₁₁NO₃
Molecular Weight: 181,19 g/mol
Schmelzpunkt: 32…33 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09038482
CAS-Nummer: 2475-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR