4,6-Dibrom-2,5-dichloranilin

Lieferant: CHEMPUR

RH01208.250 RH01208.1
CHMPRH01208.250EA 0 EUR
CHMPRH01208.250 MAYBRH01208.1
4,6-Dibrom-2,5-dichloranilin
4,6-Dibrom-2,5-dichloranilin
Formel: C₆H₃Br₂Cl₂N
CAS-Nummer: 27761-65-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR