3,5-Dichlor-2,4,6-trifluorbenzotrifluorid

Lieferant: FLUOROCHEM

010449-100G 010449-25G
FLUO010449-100GEA 0 EUR
FLUO010449-100G FLUO010449-25G
3,5-Dichlor-2,4,6-trifluorbenzotrifluorid
3,5-Dichlor-2,4,6-trifluorbenzotrifluorid
Formel: C₇Cl₂F₆
MDL: MFCD00039304
CAS-Nummer: 4284-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR