3,5-Dichlor-2-fluorbenzotrifluorid

Lieferant: Thermo Scientific

SEW04467.250 SEW04467.1
MAYBSEW04467.250EA 0 EUR
MAYBSEW04467.250 MAYBSEW04467.1
3,5-Dichlor-2-fluorbenzotrifluorid
3,5-Dichlor-2-fluorbenzotrifluorid
Formel: C₇H₂Cl₂F₄
MDL: MFCD00205816
CAS-Nummer: 258348-32-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR