3-Chlor-4,5-difluorbenzotrifluorid

Lieferant: FLUOROCHEM

004395-1G 004395-5G
FLUO004395-1GEA 155 EUR
FLUO004395-1G FLUO004395-5G
3-Chlor-4,5-difluorbenzotrifluorid
3-Chlor-4,5-difluorbenzotrifluorid
Formel: C₇H₂ClF₅
MDL: MFCD00153090
CAS-Nummer: 77227-99-7
EINECS: 401-930-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR