4'-Fluor-2-methylpropiophenon ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4'-Fluoroisobutyrophenone, 1-(4-Fluorophenyl)-2-methylpropan-1-one
H26036.03
H26036.06
H26036.03EA
138
EUR
H26036.03
H26036.06
4'-Fluor-2-methylpropiophenon ≥97%
4'-Fluor-2-methylpropiophenon
Purity: 97%
CAS No.: 26393-91-9
Molecular Formula: C10H11FO
Formula Weight: 166.19
Refractive Index: 1.4980
Formel:
C₁₀H₁₁FO Molecular Weight: 166.19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038305 CAS-Nummer: 26393-91-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...