3,5-Dinitro-4-fluorbenzotrifluorid
Lieferant: FLUOROCHEM
004967-1G
004967-5G
FLUO004967-1GEA
219
EUR
FLUO004967-1G
FLUO004967-5G
3,5-Dinitro-4-fluorbenzotrifluorid
3,5-Dinitro-4-fluorbenzotrifluorid
Formel:
C₇H₃F₃N₂O₄ |
MDL:
MFCD00153131 CAS-Nummer: 393-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...