1-Brom-2,4-bis(trifluormethyl)benzol
Lieferant: FLUOROCHEM
Synonyme:
2,4-Bis(trifluoromethyl)-1-bromobenzene
006025-5G
006025-25G
FLUO006025-5GEA
55.2
EUR
FLUO006025-5G
FLUO006025-25G
1-Brom-2,4-bis(trifluormethyl)benzol
1-Brom-2,4-bis(trifluormethyl)benzol
Formel:
C₈H₃BrF₆ Molecular Weight: 293.01 g/mol |
MDL:
MFCD00074904 CAS-Nummer: 327-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...