1-Brom-2,4-bis(trifluormethyl)benzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4-Bis(trifluoromethyl)-1-bromobenzene

M-Clarity™ Qualitätsstufe = MQ100

330388-1G 330388-5G
SIAL330388-1GEA 30.3 EUR
SIAL330388-1G SIAL330388-5G
1-Brom-2,4-bis(trifluormethyl)benzol, Sigma-Aldrich®
1-Brom-2,4-bis(trifluormethyl)benzol
Formel: C₈H₃BrF₆
Molecular Weight: 293,01 g/mol
MDL: MFCD00074904
CAS-Nummer: 327-75-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR