2,4-Dichlor-5-methylbenzotrifluorid
Lieferant: FLUOROCHEM
007842-5G
007842-25G
007842-1G
FLUO007842-5GEA
0
EUR
FLUO007842-5G
FLUO007842-25G
FLUO007842-1G
2,4-Dichlor-5-methylbenzotrifluorid
2,4-Dichlor-5-methylbenzotrifluorid
Formel:
C₈H₅Cl₂F₃ |
MDL:
MFCD00190113 CAS-Nummer: 115571-61-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...