4,6-Dichlor-2-(trifluormethyl)chinolin

Lieferant: COMBI-BLOCKS

HC-2147-5G HC-2147-1G
COBBHC-2147-5GEA 886 EUR
COBBHC-2147-5G COBBHC-2147-1G
4,6-Dichlor-2-(trifluormethyl)chinolin
4,6-Dichloro-2-(trifluoromethyl)quinoline
Formel: C₁₀H₄Cl₂F₃N
MDL: MFCD00276578
CAS-Nummer: 18706-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR