4,7-Dichlor-2-(trifluormethyl)chinolin

Lieferant: COMBI-BLOCKS

HC-2212-1G HC-2212-5G
COBBHC-2212-1GEA 404 EUR
COBBHC-2212-1G COBBHC-2212-5G
4,7-Dichlor-2-(trifluormethyl)chinolin
4,7-Dichloro-2-(trifluoromethyl)quinoline
Formel: C₁₀H₄Cl₂F₃N
MDL: MFCD11504914
CAS-Nummer: 702640-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR