4,7-Dichlor-8-fluor-2-(trifluormethyl)chinolin

Lieferant: COMBI-BLOCKS

HC-2568-1G HC-2568-5G
COBBHC-2568-1GEA 149 EUR
COBBHC-2568-1G COBBHC-2568-5G
4,7-Dichlor-8-fluor-2-(trifluormethyl)chinolin
4,7-Dichloro-8-fluoro-2-(trifluoromethyl)quinoline
Formel: C₁₀H₃Cl₂F₄N
MDL: MFCD12026031
CAS-Nummer: 1150164-86-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR