4,8-Dichlor-2-(trifluormethyl)chinolin
Lieferant: Thermo Scientific
KM09491.1
KM09491.10
MAYBKM09491.1EA
0
EUR
MAYBKM09491.1
MAYBKM09491.10
4,8-Dichlor-2-(trifluormethyl)chinolin
4,8-Dichloro-2-(trifluoromethyl)quinoline
Formel:
C₁₀H₄Cl₂F₃N Molecular Weight: 266,0467 g/mol |
MDL:
MFCD00276590 CAS-Nummer: 18706-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...