4-Chlor-7-(trifluormethyl)chinolin

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-7-trifluoromethylquinoline

HC-6560-5G HC-6560-25G HC-6560-1G
COBBHC-6560-5GEA 143 EUR
COBBHC-6560-5G COBBHC-6560-25G COBBHC-6560-1G
4-Chlor-7-(trifluormethyl)chinolin
4-Chloro-7-(trifluoromethyl)quinoline
Formel: C₁₀H₅ClF₃N
Molecular Weight: 231,6016 g/mol
MDL: MFCD00006775
CAS-Nummer: 346-55-4
EINECS: 206-471-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR