4-Chlor-8-fluor-2-(trifluormethyl)chinolin

Lieferant: CHEMPUR

KM09508.250 KM09508.1
CHMPKM09508.250EA 0 EUR
CHMPKM09508.250 MAYBKM09508.1
4-Chlor-8-fluor-2-(trifluormethyl)chinolin
4-Chloro-8-fluoro-2-(trifluoromethyl)quinoline
Formel: C₁₀H₄ClF₄N
Molecular Weight: 249,5921 g/mol
MDL: MFCD01570536
CAS-Nummer: 401567-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR