8-Chlor-6-(trifluormethyl)chinolin
Lieferant: COMBI-BLOCKS
HC-3035-5G
HC-3035-1G
COBBHC-3035-5GEA
0
EUR
COBBHC-3035-5G
COBBHC-3035-1G
8-Chlor-6-(trifluormethyl)chinolin
8-Chloro-6-(trifluoromethyl)quinoline
Formel:
C₁₀H₅ClF₃N |
MDL:
MFCD13195692 CAS-Nummer: 1215206-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...