(1-Isopropyl-4-piperidinyl)methanol

Lieferant: CHEMPUR

Synonyme: (1-Isopropylpiperidin-4-yl)methanol

60-0145.1 60-0145.5
CHMP60-0145.1EA 0 EUR
CHMP60-0145.1 CHMP60-0145.5
(1-Isopropyl-4-piperidinyl)methanol
(1-Isopropyl-4-piperidinyl)methanol
Formel: C₉H₁₉NO
Molecular Weight: 157,2533 g/mol
MDL: MFCD08059811
CAS-Nummer: 280774-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR