tert-Butyl(dimethylphosphono)acetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
tert-Butyl 2-(dimethoxyphosphoryl)acetate, Dimethylphosphonoacetic acid tert-Butyl ester, tert-Butyl dimethylphosphonoacetate, Dimethyl (tert-Butoxycarbonylmethyl)phosphonate, Dimethyl (Boc-methyl)phosphonate
79522-100ML
79522-25ML
SIAL79522-100MLEA
1010
EUR
SIAL79522-100ML
SIAL79522-25ML
tert-Butyl(dimethylphosphono)acetat, Sigma-Aldrich®
tert-Butyl(dimethylphosphono)acetat
Formel:
C₈H₁₇O₅P Molecular Weight: 224,1913 g/mol |
MDL:
MFCD00042939 CAS-Nummer: 62327-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...