Methyl(diethylphosphono)acetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diethyl methoxycarbonylmethanephosphonate, Methyl 2-(diethoxyphosphoryl)acetate, Methyl diethylphosphonoacetate

M-Clarity™ Qualitätsstufe = MQ100

158763-10G 158763-50G
SIAL158763-10GEA 78.4 EUR
SIAL158763-10G SIAL158763-50G
Methyl(diethylphosphono)acetat, Sigma-Aldrich®
Methyl(diethylphosphono)acetat
Formel: C₇H₁₅O₅P
Molecular Weight: 210,17 g/mol
MDL: MFCD00009081
CAS-Nummer: 1067-74-9
EINECS: 213-938-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR