1-(4-Fluor-2-nitrophenyl)piperidin

Lieferant: COMBI-BLOCKS

AN-1225-5G AN-1225-25G AN-1225-1G
COBBAN-1225-5GEA 127 EUR
COBBAN-1225-5G COBBAN-1225-25G COBBAN-1225-1G
1-(4-Fluor-2-nitrophenyl)piperidin
1-(4-Fluor-2-nitrophenyl)piperidin
Formel: C₁₁H₁₃FN₂O₂
Molecular Weight: 224,23 g/mol
MDL: MFCD00798370
CAS-Nummer: 719-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR