2,4-Bis(trifluormethyl)phenylacetonitril
Lieferant: FLUOROCHEM
007831-1G
007831-2.5G
FLUO007831-1GEA
82.1
EUR
FLUO007831-1G
FLUO007831-2.5G
2,4-Bis(trifluormethyl)phenylacetonitril
2,4-Bis(trifluormethyl)phenylacetonitril
Formel:
C₁₀H₅F₆N |
CAS-Nummer:
201789-28-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...