3,5-Bis(trifluormethyl)phenylacetonitril

Lieferant: FLUOROCHEM

Synonyme: 2-(3,5-Bis(trifluoromethyl)phenyl)acetonitrile

002106-1G 002106-5G 002106-25G
FLUO002106-1GEA 31.1 EUR
FLUO002106-1G FLUO002106-5G FLUO002106-25G
3,5-Bis(trifluormethyl)phenylacetonitril
3,5-Bis(trifluormethyl)phenylacetonitril
Formel: C₁₀H₅F₆N
MDL: MFCD00009904
CAS-Nummer: 85068-32-2
EINECS: 285-294-0
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR