5-Brom-1,3-benzodioxol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-1,2-(methylenedioxy)benzene, 5-Bromobenzo[d][1,3]dioxole

OS-7646-25G OS-7646-100G
COBBOS-7646-25GEA 46.2 EUR
COBBOS-7646-25G COBBOS-7646-100G
5-Brom-1,3-benzodioxol
5-Brom-1,3-benzodioxol
Formel: C₇H₅BrO₂
Molecular Weight: 201.02 g/mol
MDL: MFCD00005821
CAS-Nummer: 2635-13-4
EINECS: 220-123-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR