(2-Methylbenzyl)phosphonsäure
Lieferant: FLUOROCHEM
001310-1G
001310-5G
FLUO001310-1GEA
46.2
EUR
FLUO001310-1G
FLUO001310-5G
(2-Methylbenzyl)phosphonsäure
(2-Methylbenzyl)phosphonsäure
Formel:
C₈H₁₁O₃P |
MDL:
MFCD00041412 CAS-Nummer: 18896-56-5 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...