Ethyl-4-piperidincarboxylat

Lieferant: FLUOROCHEM

Synonyme: Ethyl piperidine-4-carboxylate, Ethyl isonipecotate, Ethyl 4-Piperidinecarboxylate

009839-25G 009839-100G
FLUO009839-25GEA 36.9 EUR
FLUO009839-25G FLUO009839-100G
Ethyl-4-piperidincarboxylat
Ethyl-4-piperidincarboxylat
Formel: C₈H₁₅NO₂
Molecular Weight: 157.21 g/mol
MDL: MFCD00006003
CAS-Nummer: 1126-09-6
EINECS: 214-416-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR