1,2-Naphthochinon-4-sulfonsäure Natriumsalz ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Folin's reagent, Sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulphonate

A10507.14 A10507.22
A10507.14EA 93.7 EUR
A10507.14 A10507.22
1,2-Naphthochinon-4-sulfonsäure Natriumsalz ≥97%
1,2-Naphthochinon-4-sulfonsäure Natriumsalz

Purity: 97% CAS No.: 521-24-4 Molecular Formula: C10H5NaO5S Formula Weight: 260.20 Melting Point: ca 289° dec. Solubility: Soluble in water. Moderately soluble in acetone. Slightly soluble in alcohol Beilstein Reference: 4169788 Merck Reference: 14,8646 MDL No.: MFCD00001700

Formel: C₁₀H₅NaO₅S
Molecular Weight: 260.20 g/mol
Schmelzpunkt: ∼ 289 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001700
CAS-Nummer: 521-24-4
EINECS: 208-308-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR