Dimethyl(2-oxopropyl)phosphonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dimethyl acetylmethylphosphonate

180696-25G 180696-5G
SIAL180696-25GEA 169 EUR
SIAL180696-25G SIAL180696-5G
Dimethyl(2-oxopropyl)phosphonat, Sigma-Aldrich®
Dimethyl(2-oxopropyl)phosphonat
Formel: C₅H₁₁O₄P
Molecular Weight: 166,12 g/mol
MDL: MFCD00008769
CAS-Nummer: 4202-14-6
EINECS: 224-110-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR