Trimethylsilyldiethylphosphonoacetat

Lieferant: CHEMPUR SILANE

SILS19250.5 SILS19250.25
SILNSILS19250.5EA 0 EUR
SILNSILS19250.5 SILNSILS19250.25
Trimethylsilyldiethylphosphonoacetat
Trimethylsilyldiethylphosphonoacetat
Formel: C₉H₂₁O₅PSi
MDL: MFCD00009080
CAS-Nummer: 66130-90-3
EINECS: 266-171-0
UN: 1993
ADR: 3,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR