Tetraethyl((dimethylamino)methylen)bisphosphonat 97%

Lieferant: Thermo Fisher Scientific
Achtung

A14581.06 A14581.14
A14581.06EA 41.9 EUR
A14581.06 A14581.14
Tetraethyl((dimethylamino)methylen)bisphosphonat 97%
Tetraethyl((dimethylamino)methylen)bisphosphonat

Purity: 97% CAS No.: 18855-52-2 Molecular Formula: C11H27NO6P2 Formula Weight: 331.29 Storage Temperature: 2°C to 8°C Refractive Index: 1.4510 Boiling Point: 138-140°/1mm Beilstein Reference: 1914890 MDL No.: MFCD00015128

Formel: C₁₁H₂₇NO₆P₂
Molecular Weight: 331,29 g/mol
Siedepunkt: 138…140 °C (1 mmHg)
MDL: MFCD00015128
CAS-Nummer: 18855-52-2
EINECS: 242-627-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR