1-(2-Brom-4-nitrophenyl)piperidin-4-ol

Lieferant: COMBI-BLOCKS

HC-2132-5G HC-2132-1G
COBBHC-2132-5GEA 0 EUR
COBBHC-2132-5G COBBHC-2132-1G
1-(2-Brom-4-nitrophenyl)piperidin-4-ol
1-(2-Brom-4-nitrophenyl)piperidin-4-ol
Formel: C₁₁H₁₃BrN₂O₃
MDL: MFCD11039803
CAS-Nummer: 1072944-61-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR