2,2-Difluor-1,3-benzodioxol-4-carbaldehyd

Lieferant: FLUOROCHEM

Synonyme: 2,2-Difluorobenzo[d][1,3]dioxole-4-carbaldehyde

007816-5G 007816-1G
FLUO007816-5GEA 267 EUR
FLUO007816-5G FLUO007816-1G
2,2-Difluor-1,3-benzodioxol-4-carbaldehyd
2,2-Difluor-1,3-benzodioxol-4-carbaldehyd
Formel: C₈H₄F₂O₃
Molecular Weight: 186.11 g/mol
MDL: MFCD00792419
CAS-Nummer: 119895-68-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR