2,2-Difluor-1,3-benzodioxol-4-carbonsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,2-Difluorobenzo[d][1,3]dioxole-4-carboxylic acid
B23955.03
B23955.06
B23955.03EA
39
EUR
B23955.03
B23955.06
2,2-Difluor-1,3-benzodioxol-4-carbonsäure ≥97%
2,2-Difluor-1,3-benzodioxol-4-carbonsäure
Purity: 97%
CAS No.: 126120-85-2
Molecular Formula: C8H4F2O4
Formula Weight: 202.12
Melting Point: 202-205°
Beilstein Reference: 7292629
Formel:
C₈H₄F₂O₄ Molecular Weight: 202,12 g/mol Schmelzpunkt: 202…205 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631473 CAS-Nummer: 126120-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...