2,2-Difluor-1,3-benzodioxol-5-carbonsäure
Lieferant: FLUOROCHEM
Synonyme:
2,2-Difluorobenzo[d][1,3]dioxole-5-carboxylic acid
007814-10G
007814-5G
007814-1G
FLUO007814-10GEA
201
EUR
FLUO007814-10G
FLUO007814-5G
FLUO007814-1G
2,2-Difluor-1,3-benzodioxol-5-carbonsäure
2,2-Difluor-1,3-benzodioxol-5-carbonsäure
Formel:
C₈H₄F₂O₄ Molecular Weight: 202,12 g/mol |
MDL:
MFCD00792417 CAS-Nummer: 656-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...