Furan-3-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(3-Furanyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan

PN-2310-5G PN-2310-1G
COBBPN-2310-5GEA 0 EUR
COBBPN-2310-5G COBBPN-2310-1G
Furan-3-boronsäurepinakolester
Furan-3-boronsäurepinakolester
Formel: C₁₀H₁₅BO₃
MDL: MFCD03453669
CAS-Nummer: 248924-59-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR