5-Methylfuran-2-boronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4,4,5,5-Tetramethyl-2-(5-methylfuran-2-yl)-1,3,2-dioxaborolane, 2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan, (5-Methylfuran-2-yl)boronic acid Pinacol ester
PN-8849-5G
PN-8849-1G
COBBPN-8849-5GEA
87
EUR
COBBPN-8849-5G
COBBPN-8849-1G
5-Methylfuran-2-boronsäurepinakolester
5-Methylfuran-2-boronsäurepinakolester
Formel:
C₁₁H₁₇BO₃ Molecular Weight: 208,06 g/mol |
MDL:
MFCD03094689 CAS-Nummer: 338998-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...