5-Methylfuran-2-boronsäure

Lieferant: FLUOROCHEM

Synonyme: 5-Methyl-2-furanboronic acid, 5-Methyl-2-furylboronic acid, (5-Methylfuran-2-yl)boronic acid

011049-5G 011049-1G
FLUO011049-5GEA 324 EUR
FLUO011049-5G FLUO011049-1G
5-Methylfuran-2-boronsäure
5-Methylfuran-2-boronsäure
Formel: C₅H₇BO₃
Molecular Weight: 125.9183 g/mol
MDL: MFCD01114644
CAS-Nummer: 62306-79-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR