2,5-Dimethylfuran-3-boronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-(2,5-Dimethylfuran-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-5503-1G
PN-5503-250M
COBBPN-5503-1GEA
1110
EUR
COBBPN-5503-1G
COBBPN-5503-250M
2,5-Dimethylfuran-3-boronsäurepinakolester
2,5-Dimethylfuran-3-boronsäurepinakolester
Formel:
C₁₂H₁₉BO₃ Molecular Weight: 222.885 g/mol |
MDL:
MFCD12405448 CAS-Nummer: 1025718-96-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...