Tri-tert-butylphosphoniumtetrafluoroborat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Tri-t-butylphosphonium tetrafluoroborate
M-Clarity™ Qualitätsstufe = MQ100
578940-1G
578940-5G
SIAL578940-1GEA
98.1
EUR
SIAL578940-1G
SIAL578940-5G
Tri-tert-butylphosphoniumtetrafluoroborat, Sigma-Aldrich®
Tri-tert-butylphosphoniumtetrafluoroborat
Formel:
C₁₂H₂₈BF₄P Molecular Weight: 290,13 g/mol |
MDL:
MFCD04039975 CAS-Nummer: 131274-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...