Chinuclidin Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Azabicyclo[2.2.2]octane hydrochloride
M-Clarity™ Qualitätsstufe = MQ100
135917-1G
SIAL135917-1GEA
114
EUR
SIAL135917-1G
Chinuclidin Hydrochlorid, Sigma-Aldrich®
Chinuclidin Hydrochlorid
Formel:
C₇H₁₄ClN Molecular Weight: 147,650 g/mol |
MDL:
MFCD00012727 CAS-Nummer: 39896-06-5 EINECS: 254-682-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...