(±)-3-Aminochinuclidin Dihydrochlorid

Lieferant: Thermo Scientific

Synonyme: 3-Aminoquinuclidine dihydrochloride, 3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride

RJC00652.1 RJC00652.10 RJC00652.50
MAYBRJC00652.1EA 0 EUR
MAYBRJC00652.1 MAYBRJC00652.10 MAYBRJC00652.50
(±)-3-Aminochinuclidin Dihydrochlorid
(±)-3-Aminochinuclidin Dihydrochlorid
Formel: C₇H₁₆Cl₂N₂
Molecular Weight: 199,1213 g/mol
MDL: MFCD00137395
CAS-Nummer: 6530-09-2
EINECS: 229-424-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR