6-Hydroxy-3-coumaranon ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Hydroxybenzofuran-3(2H)-one, 6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one

L10951.03 L10951.06
L10951.03EA 32.4 EUR
L10951.03 L10951.06
6-Hydroxy-3-coumaranon ≥97%
6-Hydroxy-3-coumaranon

Purity: 97% CAS No.: 6272-26-0 Molecular Formula: C8H6O3 Formula Weight: 150.14 Melting Point: 240-242° Beilstein Reference: 121444

Formel: C₈H₆O₃
Molecular Weight: 150.14 g/mol
Schmelzpunkt: 240…242 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00068174
CAS-Nummer: 6272-26-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR