6-Hydroxy-3-coumaranon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
6-Hydroxybenzofuran-3(2H)-one, 6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one
M-Clarity™ Qualitätsstufe = MQ100
630039-1G
630039-5G
SIAL630039-1GEA
26
EUR
SIAL630039-1G
SIAL630039-5G
6-Hydroxy-3-coumaranon, Sigma-Aldrich®
6-Hydroxy-3-coumaranon
Formel:
C₈H₆O₃ Molecular Weight: 150,14 g/mol |
MDL:
MFCD00068174 CAS-Nummer: 6272-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...