Benzo[b]furan-2-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Benzofuran-2-boronic acid, 2-Benzofurylboronic acid

BB-2653-5G BB-2653-25G
COBBBB-2653-5GEA 146 EUR
COBBBB-2653-5G COBBBB-2653-25G
Benzo[b]furan-2-boronsäure
Benzo[b]furan-2-boronsäure
Formel: C₈H₇BO₃
Molecular Weight: 161,95 g/mol
MDL: MFCD00236019
CAS-Nummer: 98437-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR