Benzo[b]furan-2-boronsäure

Lieferant: FLUOROCHEM

010978-5G 010978-1G
FLUO010978-5GEA 122 EUR
FLUO010978-5G FLUO010978-1G
Benzo[b]furan-2-boronsäure
Benzo[b]furan-2-boronsäure
Formel: C₈H₇BO₃
MDL: MFCD00236019
CAS-Nummer: 98437-24-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR