Benzo[b]furan-2-boronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Benzofuran-2-boronic acid, 2-Benzofurylboronic acid

M-Clarity™ Qualitätsstufe = MQ100

499943-25G 499943-5G
SIAL499943-25GEA 163 EUR
SIAL499943-25G SIAL499943-5G
Benzo[b]furan-2-boronsäure, Sigma-Aldrich®
Benzo[b]furan-2-boronsäure
Formel: C₈H₇BO₃
Molecular Weight: 161,95 g/mol
MDL: MFCD00236019
CAS-Nummer: 98437-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR